COM964464986 Combi-Blocks, Inc.

Sodium 8-amino-4-oxo-3,4-dihydrophthalazin-1-olate

  • MDL number MFCD14702360
  • Cas Number : 20666-12-0
  • Molecular Weight : 199.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5439

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5439
Availability details for
Quantity
  • Availability
  • Add to Cart