COM964465007 Combi-Blocks, Inc.

(R)-1-(4-Methoxyphenyl)propan-1-amine hydrochloride

  • MDL number MFCD12910568
  • Cas Number : 379711-94-1
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5501

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5501
Availability details for
Quantity
  • Availability
  • Add to Cart