COM964465019 Combi-Blocks, Inc.

1,2-Di(thiazol-2-yl)ethane-1,2-dione

  • MDL number MFCD30529676
  • Cas Number : 114698-06-5
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5527

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5527
Availability details for
Quantity
  • Availability
  • Add to Cart