COM964465049 Combi-Blocks, Inc.

2-Chloroquinazolin-8-ol

  • MDL number MFCD24728919
  • Cas Number : 953039-10-6
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5605

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5605
Availability details for
Quantity
  • Availability
  • Add to Cart