COM964465094 Combi-Blocks, Inc.

1-(2-Pyrimidinyl)-l-proline

  • MDL number MFCD08059688
  • Cas Number : 1165543-55-4
  • Molecular Weight : 193.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5727

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5727
Availability details for
Quantity
  • Availability
  • Add to Cart