COM964465098 Combi-Blocks, Inc.

3-(Carboxymethoxy)benzoic acid

  • MDL number MFCD09259431
  • Cas Number : 1878-61-1
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5736

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5736
Availability details for
Quantity
  • Availability
  • Add to Cart