COM964465107 Combi-Blocks, Inc.

2,3-Dimethyl-imidazo[1,2-a]pyridin-8-ylamine

  • MDL number MFCD16658152
  • Cas Number : 119858-51-4
  • Molecular Weight : 161.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5766
Availability details for
Quantity
  • Availability
  • Add to Cart