COM964465154 Combi-Blocks, Inc.

Methyl 3-(3-bromophenyl)-2-((tert-butoxycarbonyl)amino)-2-methylpropanoate

  • MDL number MFCD30185886
  • Cas Number : 887969-56-4
  • Molecular Weight : 372.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5888

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5888
Availability details for
Quantity
  • Availability
  • Add to Cart