COM964465184 Combi-Blocks, Inc.

Methyl 3-(4-bromothiazol-2-yl)-3-azabicyclo[3.2.1]octane-8-carboxylate

  • MDL number MFCD30472141
  • Cas Number : 2007909-42-2
  • Molecular Weight : 331.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5982

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5982
Availability details for
Quantity
  • Availability
  • Add to Cart