COM964465202 Combi-Blocks, Inc.

Methyl (r)-(benzhydrylsulfinyl)acetate

  • MDL number MFCD12031454
  • Cas Number : 713134-72-6
  • Molecular Weight : 288.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6031
Availability details for
Quantity
  • Availability
  • Add to Cart