COM964465225 Combi-Blocks, Inc.

6-Amino-5-methylpyridin-3-ylboronic acid hydrochloride

  • MDL number MFCD30471757
  • Cas Number : 2304635-84-3
  • Molecular Weight : 188.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6087

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6087
Availability details for
Quantity
  • Availability
  • Add to Cart