COM964465238 Combi-Blocks, Inc.

4-Iodonaphthalen-1-ol

  • MDL number MFCD17012478
  • Cas Number : 113855-57-5
  • Molecular Weight : 270.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6117

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6117
Availability details for
Quantity
  • Availability
  • Add to Cart