COM964465267 Combi-Blocks, Inc.

5-Bromo-n-(2-methylpyridin-4-yl)thiophene-2-carboxamide

  • MDL number MFCD22205058
  • Cas Number : 1468774-80-2
  • Molecular Weight : 297.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6187

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6187
Availability details for
Quantity
  • Availability
  • Add to Cart