COM964465280 Combi-Blocks, Inc.

N-Cyanobenzenecarboximidamide

  • MDL number MFCD03428319
  • Cas Number : 17513-09-6
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6222

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6222
Availability details for
Quantity
  • Availability
  • Add to Cart