COM964465295 Combi-Blocks, Inc.

Heptyl methyl sulfide

  • MDL number MFCD00027314
  • Cas Number : 20291-61-6
  • Molecular Weight : 146.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6252

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6252
Availability details for
Quantity
  • Availability
  • Add to Cart