COM964465433 Combi-Blocks, Inc.

4,6-Difluoro-2,3-dihydro-1h-inden-1-amine

  • MDL number MFCD07373917
  • Cas Number : 148960-40-1
  • Molecular Weight : 169.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6683

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6683
Availability details for
Quantity
  • Availability
  • Add to Cart