COM964465486 Combi-Blocks, Inc.

2,5-Dichloro-4-fluoroaniline

  • MDL number MFCD06808808
  • Cas Number : 2729-37-5
  • Molecular Weight : 180.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6814

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6814
Availability details for
Quantity
  • Availability
  • Add to Cart