COM964465498 Combi-Blocks, Inc.

Ethyl 2-[3,5-di(trifluoromethyl)anilino]-2-oxoacetate

  • MDL number MFCD00119974
  • Cas Number : 69065-93-6
  • Molecular Weight : 329.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6851

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6851
Availability details for
Quantity
  • Availability
  • Add to Cart