COM964465505 Combi-Blocks, Inc.

2-Amino-7-methoxyquinazolin-4(3h)-one

  • MDL number MFCD30291126
  • Cas Number : 181871-74-9
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6867

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6867
Availability details for
Quantity
  • Availability
  • Add to Cart