COM964465524 Combi-Blocks, Inc.

Methyl 3-(1-tritylimidazol-4-yl) propionate

  • MDL number MFCD04039625
  • Cas Number : 102676-60-8
  • Molecular Weight : 396.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6918
Availability details for
Quantity
  • Availability
  • Add to Cart