COM964465529 Combi-Blocks, Inc.

4-Bromo-5-methyl-3-(pyridin-3-yl)isoxazole

  • MDL number MFCD29921230
  • Cas Number : 1956380-27-0
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6931

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6931
Availability details for
Quantity
  • Availability
  • Add to Cart