COM964465555 Combi-Blocks, Inc.

((1S,2R,3Ar,4s,7r,7as)-2-vinyl-2,3,3a,4,7,7a-hexahydro-1h-4,7-methanoinden-1-yl)methanol

  • MDL number MFCD30230775
  • Cas Number : 77189-15-2
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7003

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7003
Availability details for
Quantity
  • Availability
  • Add to Cart