COM964465591 Combi-Blocks, Inc.

1,8-Dinitronaphthalene

  • MDL number MFCD00003918
  • Cas Number : 602-38-0
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7109

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7109
Availability details for
Quantity
  • Availability
  • Add to Cart