COM964465635 Combi-Blocks, Inc.

tert-Butyl (6-oxoheptyl)carbamate

  • MDL number MFCD24468362
  • Cas Number : 128372-98-5
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7225

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7225
Availability details for
Quantity
  • Availability
  • Add to Cart