COM964465643 Combi-Blocks, Inc.

2-(2-Bromo-5-methoxyphenyl)pyrrolidine

  • MDL number MFCD09945288
  • Cas Number : 1016818-55-5
  • Molecular Weight : 256.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7248

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7248
Availability details for
Quantity
  • Availability
  • Add to Cart