COM964465686 Combi-Blocks, Inc.

5-Hydroxy-7-methylisobenzofuran-1(3h)-one

  • MDL number MFCD27977345
  • Cas Number : 1823328-98-8
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7374

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7374
Availability details for
Quantity
  • Availability
  • Add to Cart