COM964465704 Combi-Blocks, Inc.

1H-Indazol-5-amine hydrochloride

  • MDL number MFCD06801367
  • Cas Number : 64309-76-8
  • Molecular Weight : 169.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7436

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7436
Availability details for
Quantity
  • Availability
  • Add to Cart