COM964465719 Combi-Blocks, Inc.

(3-(Piperidin-2-yl)phenyl)-carbamic acid tert-butyl ester

  • MDL number MFCD06656743
  • Cas Number : 887589-33-5
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7480

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7480
Availability details for
Quantity
  • Availability
  • Add to Cart