COM964465737 Combi-Blocks, Inc.

(S)-6-Methyl-2,3-dihydro-1h-inden-1-amine hcl

  • MDL number MFCD26585384
  • Cas Number : 1956435-48-5
  • Molecular Weight : 183.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7529

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7529
Availability details for
Quantity
  • Availability
  • Add to Cart