COM964465739 Combi-Blocks, Inc.

3-Amino-5-ethyl-4,6-dimethylthieno[2,3-b]pyridine-2-carbonitrile

  • MDL number MFCD00569887
  • Cas Number : 312535-65-2
  • Molecular Weight : 231.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7535

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7535
Availability details for
Quantity
  • Availability
  • Add to Cart