COM964465755 Combi-Blocks, Inc.

2-Acetyl-6-benzyl-n,n-dimethyl-2,6-diazaspiro[3.4]octane-8-carboxamide

  • MDL number MFCD29917114
  • Cas Number : 1956319-02-0
  • Molecular Weight : 315.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7580

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7580
Availability details for
Quantity
  • Availability
  • Add to Cart