COM964465771 Combi-Blocks, Inc.

Zinc phenolsulfonate octahydrate

  • MDL number MFCD06202320
  • Cas Number : 1300-55-6
  • Molecular Weight : 555.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7624

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7624
Availability details for
Quantity
  • Availability
  • Add to Cart