COM964465796 Combi-Blocks, Inc.

2-Chloro-n-(4-chloro-3-fluorophenyl)-5-nitropyrimidin-4-amine

  • MDL number MFCD30471874
  • Cas Number : 1625680-32-1
  • Molecular Weight : 303.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7682

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7682
Availability details for
Quantity
  • Availability
  • Add to Cart