COM964465799 Combi-Blocks, Inc.

N-(3-Fluorophenyl)-3-formyl-n-methylbenzamide

  • MDL number MFCD23104833
  • Cas Number : 168155-52-0
  • Molecular Weight : 257.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7687

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7687
Availability details for
Quantity
  • Availability
  • Add to Cart