COM964465826 Combi-Blocks, Inc.

4-Fluoro-4'-(trans-4-pentylcyclohexyl)-1,1'-biphenyl

  • MDL number MFCD30187300
  • Cas Number : 81793-59-1
  • Molecular Weight : 324.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7764

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7764
Availability details for
Quantity
  • Availability
  • Add to Cart