COM964465864 Combi-Blocks, Inc.

3-Indoxyl choline phosphate

  • MDL number MFCD04972084
  • Cas Number : 439809-44-6
  • Molecular Weight : 298.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7876

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7876
Availability details for
Quantity
  • Availability
  • Add to Cart