COM964465871 Combi-Blocks, Inc.

3-Hydroxy-2-(4-bromophenyl)-propionic acid ethyl ester

  • MDL number MFCD25967320
  • Cas Number : 797756-92-4
  • Molecular Weight : 273.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7893

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7893
Availability details for
Quantity
  • Availability
  • Add to Cart