COM964465877 Combi-Blocks, Inc.

(2S,3R)-1-(Dimethylamino)-3-(3-methoxyphenyl)-2-methylpentan-3-ol (hydrochloride)

  • MDL number MFCD28385763
  • Cas Number : 809282-45-9
  • Molecular Weight : 287.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7910

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7910
Availability details for
Quantity
  • Availability
  • Add to Cart