COM964465882 Combi-Blocks, Inc.

9-(tert-Butyl)-7,12-dihydrobenzo[2,3]azepino[4,5-b]indol-6(5h)-one

  • MDL number MFCD29917024
  • Cas Number : 1169705-95-6
  • Molecular Weight : 304.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7921

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7921
Availability details for
Quantity
  • Availability
  • Add to Cart