COM964465921 Combi-Blocks, Inc.

1-(Pyrrolidin-1-yl)propan-2-one

  • MDL number MFCD12145947
  • Cas Number : 54151-38-1
  • Molecular Weight : 127.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8017

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8017
Availability details for
Quantity
  • Availability
  • Add to Cart