COM964466019 Combi-Blocks, Inc.

Octan-3-yl 16-methylheptadecanoate

  • MDL number MFCD30471708
  • Cas Number : 2007916-07-4
  • Molecular Weight : 396.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8287

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8287
Availability details for
Quantity
  • Availability
  • Add to Cart