COM964466053 Combi-Blocks, Inc.

7-Methylchroman-4-one hcl

  • MDL number MFCD30530514
  • Cas Number : 2055840-48-5
  • Molecular Weight : 198.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8366

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8366
Availability details for
Quantity
  • Availability
  • Add to Cart