COM964466075 Combi-Blocks, Inc.

(4-Methyl-pyridin-3-ylmethyl)-carbamic acid tert-butyl ester

  • MDL number MFCD28145835
  • Cas Number : 1595032-33-9
  • Molecular Weight : 222.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8441

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8441
Availability details for
Quantity
  • Availability
  • Add to Cart