COM964466084 Combi-Blocks, Inc.

3,5-Dibromo-2-ethylpyridine

  • MDL number MFCD18257696
  • Cas Number : 142337-93-7
  • Molecular Weight : 264.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8457

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8457
Availability details for
Quantity
  • Availability
  • Add to Cart