COM964466103 Combi-Blocks, Inc.

(R)-N,2-Dimethyl-n-phenylpropane-2-sulfinamide

  • MDL number MFCD30471429
  • Cas Number : 1372133-78-2
  • Molecular Weight : 211.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8518

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8518
Availability details for
Quantity
  • Availability
  • Add to Cart