COM964466123 Combi-Blocks, Inc.

7-(Methoxycarbonyl)-6,7-dihydro-5h-pyrrolizine-1-carboxylic acid

  • MDL number MFCD09701966
  • Cas Number : 92992-17-1
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8587

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8587
Availability details for
Quantity
  • Availability
  • Add to Cart