COM964466129 Combi-Blocks, Inc.

2-Amino-6-bromo-8-nitro-4(3h)-quinazolinone

  • MDL number MFCD30471467
  • Cas Number : 130148-54-8
  • Molecular Weight : 285.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8606

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8606
Availability details for
Quantity
  • Availability
  • Add to Cart