COM964466151 Combi-Blocks, Inc.

Gly-ala-gly-oh hcl

  • MDL number MFCD28385802
  • Cas Number : 143485-62-5
  • Molecular Weight : 239.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8670

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8670
Availability details for
Quantity
  • Availability
  • Add to Cart