COM964466158 Combi-Blocks, Inc.

1,6,6-Trimethyl-4,5,6,7-tetrahydro-1h-indazole-3-carboxylic acid

  • MDL number MFCD24481536
  • Cas Number : 1233243-59-8
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8686

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8686
Availability details for
Quantity
  • Availability
  • Add to Cart