COM964466205 Combi-Blocks, Inc.

Benzo[f]quinazolin-1(2h)-one, 3-amino-

  • MDL number MFCD24562106
  • Cas Number : 7066-25-3
  • Molecular Weight : 211.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8864

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8864
Availability details for
Quantity
  • Availability
  • Add to Cart